Assay Details
Gene Symbol: MYB86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343972.1 | NP_001332393.1 | 1 - 2 | 605 | 62 | |
NM_180548.4 | NP_850879.1 | 2 - 3 | 501 | 62 | ||
GenBank mRNA | AY099777.1 | - | 2 - 3 | 387 | 62 | |
AY519629.1 | - | 2 - 3 | 261 | 62 | ||
BT008740.1 | - | 2 - 3 | 261 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
