Assay Details
Gene Symbol: AT5G04170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120499.4 | NP_196037.2 | 1 - 2 | 867 | 149 | |
GenBank mRNA | AY081342.1 | - | 1 - 2 | 745 | 149 | |
BT008838.1 | - | 1 - 2 | 719 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
