Assay Details
Gene Symbol: AT5G04170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120499.4 | NP_196037.2 | 2 - 3 | 913 | 113 | |
GenBank mRNA | AY081342.1 | - | 2 - 3 | 791 | 113 | |
BT008838.1 | - | 2 - 3 | 765 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
