Assay Details
Gene Symbol: SRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120765.3 | NP_196300.1 | 9 - 10 | 984 | 83 | |
GenBank mRNA | AY518287.1 | - | 9 - 10 | 831 | 83 | |
FJ708770.1 | - | 9 - 10 | 706 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
