Assay Details
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120821.2 | NP_196356.1 | 11 - 12 | 2780 | 72 | |
GenBank mRNA | BT003857.1 | - | 11 - 12 | 2780 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
