Assay Details
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203321.1 | NP_001190250.1 | 1 - 2 | 680 | 117 | |
NM_120821.2 | NP_196356.1 | 1 - 2 | 725 | 117 | ||
GenBank mRNA | BT003857.1 | - | 1 - 2 | 725 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
