Assay Details
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203321.1 | NP_001190250.1 | 7 - 8 | 1716 | 64 | |
NM_120821.2 | NP_196356.1 | 6 - 7 | 1857 | 64 | ||
GenBank mRNA | BT003857.1 | - | 6 - 7 | 1857 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
