Assay Details
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203321.1 | NP_001190250.1 | 9 - 10 | 1921 | 110 | |
NM_120821.2 | NP_196356.1 | 8 - 9 | 2062 | 110 | ||
GenBank mRNA | BT003857.1 | - | 8 - 9 | 2062 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
