Assay Details
Gene Symbol: MPB2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342982.1 | NP_001332071.1 | 3 - 4 | 870 | 103 | |
NM_120894.5 | NP_196429.1 | 3 - 4 | 870 | 103 | ||
GenBank mRNA | AF370224.1 | - | 3 - 4 | 739 | 103 | |
AY117260.1 | - | 3 - 4 | 613 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
