Assay Details
Gene Symbol: MPB2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120894.5 | NP_196429.1 | 4 - 5 | 1081 | 98 | |
GenBank mRNA | AF370224.1 | - | 4 - 5 | 950 | 98 | |
AY117260.1 | - | 4 - 5 | 824 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
