Assay Details
Gene Symbol: CHR34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335753.1 | NP_001325209.1 | 3 - 4 | 777 | 89 | |
NM_001335754.1 | NP_001325208.1 | 1 - 2 | 244 | 89 | ||
NM_127717.2 | NP_179740.1 | 2 - 3 | 504 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
