Assay Details
Gene Symbol: AT2G22420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127806.4 | NP_179828.1 | 1 - 2 | 519 | 70 | |
GenBank mRNA | BT004021.1 | - | 1 - 2 | 233 | 70 | |
BT005050.1 | - | 1 - 2 | 202 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
