Assay Details
Gene Symbol: MES6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335853.1 | NP_001318274.1 | 2 - 3 | 518 | 79 | |
NM_127919.4 | NP_179936.3 | 2 - 3 | 536 | 79 | ||
NM_179711.2 | NP_850042.2 | 3 - 4 | 431 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
