Assay Details
Gene Symbol: MES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127925.3 | NP_179942.1 | 1 - 2 | 953 | 68 | |
GenBank mRNA | BT014901.1 | - | 1 - 2 | 381 | 68 | |
BT015031.1 | - | 1 - 2 | 374 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
