Assay Details
Gene Symbol: MES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127926.3 | NP_179943.1 | 1 - 2 | 544 | 65 | |
GenBank mRNA | BT014881.1 | - | 1 - 2 | 379 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
