Assay Details
Gene Symbol: AT5G08450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036780.1 | NP_001031857.1 | 4 - 5 | 2028 | 95 | |
NM_120930.5 | NP_196462.2 | 4 - 5 | 2106 | 95 | ||
NM_180702.3 | NP_851033.1 | 4 - 5 | 2069 | 95 | ||
GenBank mRNA | AY091073.1 | - | 4 - 5 | 1936 | 95 | |
AY122935.1 | - | 3 - 4 | 1851 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
