Assay Details
Gene Symbol: AT5G08450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036780.1 | NP_001031857.1 | 5 - 6 | 2088 | 114 | |
NM_120930.5 | NP_196462.2 | 5 - 6 | 2166 | 114 | ||
NM_180702.3 | NP_851033.1 | 5 - 6 | 2129 | 114 | ||
GenBank mRNA | AY091073.1 | - | 5 - 6 | 1996 | 114 | |
AY122935.1 | - | 4 - 5 | 1911 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
