Assay Details
Gene Symbol: AT5G08450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036780.1 | NP_001031857.1 | 7 - 8 | 2289 | 75 | |
NM_120930.5 | NP_196462.2 | 7 - 8 | 2367 | 75 | ||
NM_180702.3 | NP_851033.1 | 7 - 8 | 2330 | 75 | ||
GenBank mRNA | AY091073.1 | - | 7 - 8 | 2197 | 75 | |
AY122935.1 | - | 6 - 7 | 2112 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
