Assay Details
Gene Symbol: AT5G08450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036780.1 | NP_001031857.1 | 9 - 10 | 2459 | 126 | |
NM_120930.5 | NP_196462.2 | 9 - 10 | 2537 | 126 | ||
NM_180702.3 | NP_851033.1 | 9 - 10 | 2500 | 126 | ||
GenBank mRNA | AY091073.1 | - | 9 - 10 | 2367 | 126 | |
AY122935.1 | - | 8 - 9 | 2282 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
