Assay Details
Gene Symbol: AO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122105.3 | NP_568407.2 | 6 - 7 | 3591 | 88 | |
NM_180718.2 | NP_851049.1 | 6 - 7 | 3823 | 88 | ||
GenBank mRNA | AB005804.1 | - | 6 - 7 | 3540 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
