Assay Details
Gene Symbol: AO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122105.3 | NP_568407.2 | 9 - 10 | 3980 | 75 | |
NM_180718.2 | NP_851049.1 | 9 - 10 | 4212 | 75 | ||
GenBank mRNA | AB005804.1 | - | 9 - 10 | 3929 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
