Assay Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122128.3 | NP_568413.1 | 13 - 14 | 1484 | 83 | |
GenBank mRNA | AF242580.1 | - | 13 - 14 | 1501 | 83 | |
AY136479.1 | - | 13 - 14 | 1558 | 83 | ||
BT006610.1 | - | 13 - 14 | 1270 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
