Assay Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036844.3 | NP_001031921.3 | 1 - 2 | 397 | 116 | |
NM_122128.3 | NP_568413.1 | 1 - 2 | 397 | 116 | ||
GenBank mRNA | AF242580.1 | - | 1 - 2 | 414 | 116 | |
AY136479.1 | - | 1 - 2 | 471 | 116 | ||
BT006610.1 | - | 1 - 2 | 183 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
