Assay Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036844.3 | NP_001031921.3 | 4 - 5 | 697 | 133 | |
NM_122128.3 | NP_568413.1 | 4 - 5 | 697 | 133 | ||
GenBank mRNA | AF242580.1 | - | 4 - 5 | 714 | 133 | |
AY136479.1 | - | 4 - 5 | 771 | 133 | ||
BT006610.1 | - | 4 - 5 | 483 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
