Assay Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036844.3 | NP_001031921.3 | 6 - 7 | 846 | 134 | |
NM_122128.3 | NP_568413.1 | 6 - 7 | 846 | 134 | ||
GenBank mRNA | AF242580.1 | - | 6 - 7 | 863 | 134 | |
AY136479.1 | - | 6 - 7 | 920 | 134 | ||
BT006610.1 | - | 6 - 7 | 632 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
