Assay Details
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036844.3 | NP_001031921.3 | 8 - 9 | 948 | 137 | |
NM_122128.3 | NP_568413.1 | 8 - 9 | 948 | 137 | ||
GenBank mRNA | AF242580.1 | - | 8 - 9 | 965 | 137 | |
AY136479.1 | - | 8 - 9 | 1022 | 137 | ||
BT006610.1 | - | 8 - 9 | 734 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
