Assay Details
Gene Symbol: ACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124198.4 | NP_568694.2 | 10 - 11 | 1200 | 97 | |
NM_180823.3 | NP_851154.1 | 9 - 10 | 1270 | 97 | ||
GenBank mRNA | AF364059.1 | - | 10 - 11 | 1069 | 97 | |
AK317237.1 | - | 10 - 11 | 973 | 97 | ||
AY059736.1 | - | 8 - 9 | 938 | 97 | ||
AY091271.1 | - | 8 - 9 | 867 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
