Assay Details
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 31 - 32 | 3136 | 85 | |
NM_125093.6 | NP_200521.3 | 31 - 32 | 3185 | 85 | ||
NM_180869.3 | NP_851200.1 | 31 - 32 | 3027 | 85 | ||
GenBank mRNA | AJ249352.1 | - | 31 - 32 | 3029 | 85 | |
AK226961.1 | - | 30 - 31 | 2951 | 85 | ||
AY069869.1 | - | 31 - 32 | 3029 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
