Assay Details
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 3 - 4 | 497 | 74 | |
NM_125093.6 | NP_200521.3 | 3 - 4 | 546 | 74 | ||
NM_180869.3 | NP_851200.1 | 3 - 4 | 388 | 74 | ||
GenBank mRNA | AJ249352.1 | - | 3 - 4 | 390 | 74 | |
AK226961.1 | - | 2 - 3 | 312 | 74 | ||
AY069869.1 | - | 3 - 4 | 390 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
