Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 11 - 12 | 1426 | 79 | |
GenBank mRNA | AB017643.1 | - | 11 - 12 | 1132 | 79 | |
AY087793.1 | - | 11 - 12 | 1151 | 79 | ||
AY094441.1 | - | 11 - 12 | 1248 | 79 | ||
AY125536.1 | - | 11 - 12 | 1093 | 79 | ||
U72505.1 | - | 11 - 12 | 1118 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
