Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 2 - 3 | 464 | 76 | |
GenBank mRNA | AB017643.1 | - | 2 - 3 | 170 | 76 | |
AY087793.1 | - | 2 - 3 | 189 | 76 | ||
AY094441.1 | - | 2 - 3 | 286 | 76 | ||
AY125536.1 | - | 2 - 3 | 131 | 76 | ||
U72505.1 | - | 2 - 3 | 156 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
