Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 4 - 5 | 643 | 113 | |
GenBank mRNA | AB017643.1 | - | 4 - 5 | 349 | 113 | |
AY087793.1 | - | 4 - 5 | 368 | 113 | ||
AY094441.1 | - | 4 - 5 | 465 | 113 | ||
AY125536.1 | - | 4 - 5 | 310 | 113 | ||
U72505.1 | - | 4 - 5 | 335 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
