Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 5 - 6 | 775 | 90 | |
GenBank mRNA | AB017643.1 | - | 5 - 6 | 481 | 90 | |
AY087793.1 | - | 5 - 6 | 500 | 90 | ||
AY094441.1 | - | 5 - 6 | 597 | 90 | ||
AY125536.1 | - | 5 - 6 | 442 | 90 | ||
U72505.1 | - | 5 - 6 | 467 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
