Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 6 - 7 | 849 | 91 | |
GenBank mRNA | AB017643.1 | - | 6 - 7 | 555 | 91 | |
AY087793.1 | - | 6 - 7 | 574 | 91 | ||
AY094441.1 | - | 6 - 7 | 671 | 91 | ||
AY125536.1 | - | 6 - 7 | 516 | 91 | ||
U72505.1 | - | 6 - 7 | 541 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
