Assay Details
Gene Symbol: UPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035773.2 | NP_001030850.1 | 2 - 3 | 242 | 67 | |
NM_115169.3 | NP_190877.1 | 2 - 3 | 245 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
