Assay Details
Gene Symbol: AT3G53100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115170.5 | NP_190878.2 | 3 - 4 | 1048 | 70 | |
GenBank mRNA | AK229079.1 | - | 3 - 4 | 646 | 70 | |
BT033018.1 | - | 3 - 4 | 606 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
