Assay Details
Gene Symbol: LOG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115205.2 | NP_190913.1 | 5 - 6 | 656 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
