Assay Details
Gene Symbol: EDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339835.1 | NP_001327450.1 | 3 - 4 | 513 | 67 | |
NM_001339836.1 | NP_001327449.1 | 3 - 4 | 485 | 67 | ||
NM_115558.6 | NP_191258.2 | 3 - 4 | 513 | 67 | ||
GenBank mRNA | AY069919.1 | - | 3 - 4 | 400 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
