Assay Details
Gene Symbol: EDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339835.1 | NP_001327450.1 | 7 - 8 | 888 | 85 | |
NM_001339836.1 | NP_001327449.1 | 7 - 8 | 860 | 85 | ||
NM_115558.6 | NP_191258.2 | 7 - 8 | 888 | 85 | ||
GenBank mRNA | AY069919.1 | - | 7 - 8 | 775 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
