Assay Details
Gene Symbol: AT3G57220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115582.3 | NP_191281.1 | 9 - 10 | 1217 | 71 | |
GenBank mRNA | AK175168.1 | - | 9 - 10 | 1157 | 71 | |
AY088931.1 | - | 9 - 10 | 1153 | 71 | ||
BT029005.1 | - | 9 - 10 | 1096 | 71 | ||
KY906095.1 | - | 9 - 10 | 1096 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
