Assay Details
Gene Symbol: DEG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123346.5 | NP_568575.1 | 12 - 13 | 1337 | 101 | |
NM_203134.2 | NP_974863.1 | 12 - 13 | 1262 | 101 | ||
GenBank mRNA | AY056381.1 | - | 12 - 13 | 1281 | 101 | |
AY090319.1 | - | 12 - 13 | 1218 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
