Assay Details
Gene Symbol: DEG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123346.5 | NP_568575.1 | 8 - 9 | 997 | 91 | |
NM_203134.2 | NP_974863.1 | 8 - 9 | 964 | 91 | ||
GenBank mRNA | AY056381.1 | - | 8 - 9 | 941 | 91 | |
AY090319.1 | - | 8 - 9 | 878 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
