Assay Details
Gene Symbol: SAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198200.2 | NP_001185129.1 | 16 - 17 | 3292 | 94 | |
NM_001333042.1 | NP_001319133.1 | 16 - 17 | 3280 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
