Assay Details
Gene Symbol: ATE2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198237.1 | NP_001185166.1 | 2 - 3 | 593 | 138 | |
NM_103684.3 | NP_175222.1 | 2 - 3 | 656 | 138 | ||
GenBank mRNA | AF242581.1 | - | 2 - 3 | 455 | 138 | |
AJ417834.1 | - | 2 - 3 | 429 | 138 | ||
AY045636.1 | - | 2 - 3 | 516 | 138 | ||
AY058231.1 | - | 2 - 3 | 416 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
