Assay Details
Gene Symbol: AT1G48170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103713.3 | NP_564522.1 | 4 - 5 | 434 | 112 | |
GenBank mRNA | AK228677.1 | - | 4 - 5 | 411 | 112 | |
AY070372.1 | - | 4 - 5 | 410 | 112 | ||
AY086896.1 | - | 4 - 5 | 411 | 112 | ||
AY133765.1 | - | 4 - 5 | 387 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
