Assay Details
Gene Symbol: DCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203375.1 | NP_001190304.1 | 3 - 4 | 603 | 64 | |
NM_121360.4 | NP_196861.2 | 3 - 4 | 640 | 64 | ||
GenBank mRNA | AK119112.1 | - | 3 - 4 | 602 | 64 | |
BT005332.1 | - | 3 - 4 | 370 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
