Assay Details
Gene Symbol: PDAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343283.1 | NP_001331240.1 | 5 - 6 | 2303 | 143 | |
NM_001343284.1 | NP_001331241.1 | 5 - 6 | 1512 | 143 | ||
NM_121367.3 | NP_196868.1 | 5 - 6 | 2116 | 143 | ||
GenBank mRNA | AY052715.1 | - | 5 - 6 | 1583 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
