Assay Details
Gene Symbol: MYB40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343333.1 | NP_001330910.1 | 1 - 2 | 304 | 117 | |
NM_121438.3 | NP_196938.2 | 1 - 2 | 304 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001343333.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
