Assay Details
Gene Symbol: MYB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085120.1 | NP_001078589.1 | 2 - 3 | 486 | 75 | |
NM_001343397.1 | NP_001330734.1 | 2 - 3 | 486 | 75 | ||
NM_121535.4 | NP_197035.1 | 2 - 3 | 486 | 75 | ||
GenBank mRNA | AF370613.1 | - | 2 - 3 | 263 | 75 | |
AK228474.1 | - | 2 - 3 | 413 | 75 | ||
AY519624.1 | - | 2 - 3 | 263 | 75 | ||
BT028929.1 | - | 2 - 3 | 263 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
