Assay Details
Gene Symbol: GLIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343418.1 | NP_001332582.1 | 2 - 3 | 667 | 96 | |
NM_001343419.1 | NP_001332581.1 | 1 - 2 | 746 | 96 | ||
NM_121576.2 | NP_568318.1 | 2 - 3 | 694 | 96 | ||
GenBank mRNA | AY084802.1 | - | 2 - 3 | 389 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
